Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals
Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals

Widley used as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2 x 1 kg
Catalog number 012857.36
also known as 012857-36
Price (USD)/ Each
572.65
Online exclusive
636.00 
Save 63.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
572.65
Online exclusive
636.00 
Save 63.35 (10%)
Add to cart
Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals
Catalog number012857.36
Price (USD)/ Each
572.65
Online exclusive
636.00 
Save 63.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7704-98-5
IUPAC Nametitanium(2+) dihydride
Molecular FormulaH2Ti
InChI KeyKAZWGWWZKAHTKC-UHFFFAOYSA-N
SMILES[H-].[H-].[Ti++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
FormPowder
Assay from Suppliers CofATotal metal impurities: ≤1.00% (ICP)
Particle Size-325mesh
Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only