Thermo Scientific Chemicals

Platinum Thinfoil, 0.004^+0.0025mm (0.00016^+0.0001in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 012868.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Platinum Thinfoil, 0.004^+0.0025mm (0.00016^+0.0001in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 012868.FI
50 x 50 mm, Each
Quantity
Catalog number: 012868.FI
also known as 012868-FI
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-06-4
IUPAC Name
platinum
Molecular Formula
Pt
InChI Key
BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES
[Pt]
Form
Foil, 0.004mm ?0.001mm (0.00016 ?0.00004in) thick.
Assay from Supplier's CofA
99.95%
Assay from Supplier's CofA
Typical Analysis Ca = 1ppm
Assay from Supplier's CofA
Typical Analysis Ir = 10ppm
Assay from Supplier's CofA
Typical Analysis Na = 1ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text