Ammonium hexachloroplatinate(IV), Pt 43.4% min, Thermo Scientific Chemicals
Ammonium hexachloroplatinate(IV), Pt 43.4% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hexachloroplatinate(IV), Pt 43.4% min, Thermo Scientific Chemicals

Electroplating of platinum, manufacturing of platinum sponge
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 012882.06
also known as 012882-06
Price (USD)/ Each
579.65
Online exclusive
644.00 
Save 64.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
579.65
Online exclusive
644.00 
Save 64.35 (10%)
Add to cart
Ammonium hexachloroplatinate(IV), Pt 43.4% min, Thermo Scientific Chemicals
Catalog number012882.06
Price (USD)/ Each
579.65
Online exclusive
644.00 
Save 64.35 (10%)
-
Add to cart
Chemical Identifiers
CAS16919-58-7
IUPAC Nameplatinum(4+) diammonium hexachloride
Molecular FormulaCl6H8N2Pt
InChI KeyPCCGQTHFYHJATL-UHFFFAOYSA-J
SMILES[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 43.4% min
Electroplating of platinum, manufacturing of platinum spongeAmmonium hexachloroplatinate(IV) is used in electroplating of platinum and to make spongy platinum. It reacts with copper(II) acetate and hydrogen sulfide to prepare Cu2PtIIPt3IVS8 which finds application in diamagnetic materials and semiconductors. It is also used in platinum plating. Further, it acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only