Thermo Scientific Chemicals

Cesium nitrate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 012884.18
50 g, Each
Thermo Scientific Chemicals

Cesium nitrate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 012884.18
50 g, Each
Quantity
Catalog number: 012884.18
also known as 012884-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-18-6
IUPAC Name
caesium(1+) nitrate
Molecular Formula
CsNO3
InChI Key
NLSCHDZTHVNDCP-UHFFFAOYSA-N
SMILES
[Cs+].[O-][N+]([O-])=O
Total Metal Impurities
0.2% max.

Description

Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text