Cesium chloride, 99% (metals basis), Thermo Scientific Chemicals
Cesium chloride, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium chloride, 99% (metals basis), Thermo Scientific Chemicals

For Evacuation of Radio Tubes, Manufacturing of Cesium Compounds, Brewing
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012886.18
also known as 012886-18
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
Add to cart
Cesium chloride, 99% (metals basis), Thermo Scientific Chemicals
Catalog number012886.18
Price (USD)/ Each
78.65
Online exclusive
87.40 
Save 8.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Evacuation of radio tubes, manufacturing of cesium compounds, brewing, x-ray fluorescent screens, radiographic contrast medium.Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Evacuation of radio tubes, manufacturing of cesium compounds, brewing, x-ray fluorescent screens, radiographic contrast medium
RUO – Research Use Only