Thermo Scientific Chemicals

Cesium carbonate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012887.18
50 g, Each
Thermo Scientific Chemicals

Cesium carbonate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012887.18
50 g, Each
Quantity
Catalog number: 012887.18
also known as 012887-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-17-8
IUPAC Name
dicaesium(1+) carbonate
Molecular Formula
CCs2O3
InChI Key
FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES
[Cs+].[Cs+].[O-]C([O-])=O
Total Metal Impurities
1% max.

Description

Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text