Thermo Scientific Chemicals

Rubidium nitrate, 99%, Thermo Scientific Chemicals

Catalog number: 012889.30
250 g, Each
Thermo Scientific Chemicals

Rubidium nitrate, 99%, Thermo Scientific Chemicals

Catalog number: 012889.30
250 g, Each
Quantity
Catalog number: 012889.30
also known as 012889-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13126-12-0
IUPAC Name
rubidium(1+) nitrate
Molecular Formula
NO3Rb
InChI Key
RTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES
[Rb+].[O-][N+]([O-])=O
Form
Crystals or crystalline solid
Appearance (Color)
White
Assay from Suppliers CofA
≥98.5%

Description

Rubidium nitrate used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters. It is rarely used in fireworks to produce a red-violet colour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text