Thermo Scientific Chemicals

Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012892.22
100 g, Each
Thermo Scientific Chemicals

Rubidium chloride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012892.22
100 g, Each
Quantity
Catalog number: 012892.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-11-9
IUPAC Name
rubidium(1+) chloride
Molecular Formula
ClRb
InChI Key
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES
[Cl-].[Rb+]
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Total Metal Impurities
≤1.0%
Appearance (Color)
White
Form
Powder or crystals or crystalline powder

Description

It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text