Samarium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Samarium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
50 g
Catalog number 012895.09
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
Add to cart
Samarium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012895.09
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-19-9
IUPAC Namesamarium
Molecular FormulaSm
InChI KeyKZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES[Sm]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only