Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
5 x 100 g
Catalog number 012898.06
also known as 012898-06
Price (USD)/ Each
38.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.60
Add to cart
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012898.06
Price (USD)/ Each
38.60
-
Add to cart
Chemical Identifiers
CAS13494-98-9
IUPAC Nameyttrium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Y
InChI KeyQBAZWXKSCUESGU-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline
Total Rare Earth Oxide Impurities≤0.1% (1000 ppm)
It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only