Thermo Scientific Chemicals

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012898.22
100 g, Each
Thermo Scientific Chemicals

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012898.22
100 g, Each
Quantity
Catalog number: 012898.22
also known as 012898-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13494-98-9
IUPAC Name
yttrium(3+) hexahydrate trinitrate
Molecular Formula
H12N3O15Y
InChI Key
QBAZWXKSCUESGU-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Form
Crystalline
Total Rare Earth Oxide Impurities
≤0.1% (1000 ppm)

Description

It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text