Thermo Scientific Chemicals

Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012906.14
25 g, Each
Thermo Scientific Chemicals

Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012906.14
25 g, Each
Quantity
Catalog number: 012906.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13759-83-6
IUPAC Name
samarium(3+) hexahydrate trinitrate
Molecular Formula
H12N3O15Sm
InChI Key
HDCOFJGRHQAIPE-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
≤0.1% (1000 ppm)
Appearance (Color)
Yellow
Form
Crystalline powder or granules
Assay (special)
38.5-40.1% Sm₂O₃

Description

It is used in NMR spectroscopy for spectra resolution and is used for doping purpose. It is also used as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text