Thermo Scientific Chemicals

Praseodymium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012909.09
10 g, Each
Thermo Scientific Chemicals

Praseodymium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 012909.09
10 g, Each
Quantity
Catalog number: 012909.09
also known as 012909-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15878-77-0
IUPAC Name
praseodymium(3+) hexahydrate trinitrate
Molecular Formula
H12N3O15Pr
InChI Key
LXXCECZPOWZKLC-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Pale green
Form
Crystalline
Assay (special)
38.3-40.0% Pr₆O{11}
Total Rare Earth Oxide Impurities
≤0.1% (1000 ppm)

Description

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text