Neodymium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Neodymium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 012912.30
also known as 012912-30
Price (USD)/ Each
224.65
Online exclusive
249.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
224.65
Online exclusive
249.00 
Save 24.35 (10%)
Add to cart
Neodymium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012912.30
Price (USD)/ Each
224.65
Online exclusive
249.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS16454-60-7
IUPAC Nameneodymium(3+) hexahydrate trinitrate
Molecular FormulaH12N3NdO15
InChI KeyVQVDTKCSDUNYBO-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to purple
Assay (special)37.6-39.2% Nd₂O₃
FormCrystalline
Total Rare Earth Oxide Impurities≤0.1% (1000 ppm)
Neodymium(III) nitrate hexahydrate is used to prepare nanoparticles of neodymium oxybromide and neodymium oxide. It is also used in coloring glasses and for doping some glass lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only