Thermo Scientific Chemicals

Lanthanum(III) chloride heptahydrate, ACS, Thermo Scientific Chemicals

Catalog number: 012916.30
250 g, Each
Thermo Scientific Chemicals

Lanthanum(III) chloride heptahydrate, ACS, Thermo Scientific Chemicals

Catalog number: 012916.30
250 g, Each
Quantity
Catalog number: 012916.30
also known as 012916-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-84-0
IUPAC Name
lanthanum(3+) heptahydrate trichloride
Molecular Formula
Cl3H14LaO7
InChI Key
FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
Form
Crystals or crystalline powder
Assay from Supplier's CofA
64.5-70.0% LaCl₃
Impurity content
Calcium (Ca): ≤10ppm
Impurity content
Magnesium (Mg): ≤10ppm
Insoluble material
≤0.01%

Description

Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text