Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals
Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 012917.22
also known as 012917-22
Price (USD)/ Each
117.65
Online exclusive
138.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
138.00 
Save 20.35 (15%)
Add to cart
Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals
Catalog number012917.22
Price (USD)/ Each
117.65
Online exclusive
138.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS94219-55-3
IUPAC Namegadolinium(3+) trinitrate
Molecular FormulaGdN3O9
InChI KeyMWFSXYMZCVAQCC-UHFFFAOYSA-N
SMILES[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities≤0.5%
FormPowder or crystals or crystalline powder
It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only