Dysprosium(III) nitrate pentahydrate, 99.9% (REO), Thermo Scientific Chemicals
Dysprosium(III) nitrate pentahydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium(III) nitrate pentahydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012922.14
also known as 012922-14
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
Add to cart
Dysprosium(III) nitrate pentahydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012922.14
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Chemical Identifiers
CAS10031-49-9
IUPAC Namedysprosium(3+) pentahydrate trinitrate
Molecular FormulaDyH10N3O14
InChI KeyNOTQUFQJAWMLCE-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Dysprosium(III) nitrate pentahydrate is used to prepare carboxylate-based dinuclear dysprosium compound by reacting with n-butyric acid and triethylamine using methanol as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only