Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012928.18
also known as 012928-18
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
Add to cart
Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number012928.18
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7787-69-1
IUPAC Namecaesium(1+) bromide
Molecular FormulaBrCs
InChI KeyLYQFWZFBNBDLEO-UHFFFAOYSA-M
SMILES[Br-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cesium bromide is applied to make radiation detectors and other measuring devices. Cesium bromide has limited application in the deep IR. Cesium bromide is slightly more amenable to optical working than CsI and is sometimes utilized as a beam splitter component in wideband spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only