Thermo Scientific Chemicals

Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012929.22
100 g, Each
Thermo Scientific Chemicals

Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012929.22
100 g, Each
Quantity
Catalog number: 012929.22
also known as 012929-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3396-11-0
IUPAC Name
caesium(1+) acetate
Molecular Formula
C2H3CsO2
InChI Key
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES
[Cs+].CC([O-])=O
Total Metal Impurities
0.1% max.

Description

Cesium Acetate is used in organic synthesis especially for Perkin reaction; formation of unsaturated cinnamic-type acids by the condensation of aromatic aldehydes with fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text