Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals
Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012932.09
also known as 012932-09
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
Add to cart
Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number012932.09
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1303-11-3
IUPAC Nameindiganylidynearsane
Molecular FormulaAsIn
InChI KeyRPQDHPTXJYYUPQ-UHFFFAOYSA-N
SMILES[As]#[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
Indium arsenide is used for construction of infrared detectors, for the wavelength range of 1-3μm. It is widely used as terahertz radiation source as it is a strong photo-Dember emitter. Indium arsenide is also used for making of diode laser.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only