Thermo Scientific Chemicals

Cadmium perchlorate hexahydrate, Thermo Scientific Chemicals

Catalog number: 012936.30
250 g, Each
Thermo Scientific Chemicals

Cadmium perchlorate hexahydrate, Thermo Scientific Chemicals

Catalog number: 012936.30
250 g, Each
Quantity
Catalog number: 012936.30
also known as 012936-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10326-28-0
IUPAC Name
cadmium(2+) hexahydrate diperchlorate
Molecular Formula
CdCl2H12O14
InChI Key
PQJZOTJHMRJOEM-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cd++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
Cd 25.5-27.0%

Description

It is employed as intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text