Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals
Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012964.06
also known as 012964-06
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
28.60 
Save 3.95 (14%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
28.60 
Save 3.95 (14%)
Add to cart
Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals
Catalog number012964.06
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
28.60 
Save 3.95 (14%)
-
Add to cart
Chemical Identifiers
CAS7681-49-4
IUPAC Namesodium fluoride
Molecular FormulaFNa
InChI KeyPUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES[F-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding K0.005% max.
Impurity contentK ≈60ppm
It is used as a steel degreasing agent and a flux. It is used in heat treating salt compositions, tin-electroplating, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It has significant applications in organic synthesis including its halide-exchange with acyl chloride, sulfur chlorides, and phosphorus chloride, desilylation of silyl-protected OH groups, synthesis of fluorocarbons (Finkelstein reaction), and also in metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only