Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012967.14
25 g, Each
Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012967.14
25 g, Each
Quantity
Catalog number: 012967.14
also known as 012967-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-63-0
IUPAC Name
λ²-iron(2+) heptahydrate sulfate
Molecular Formula
FeH14O11S
InChI Key
SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
Total Metal Impurities
0.001% max.

Description

Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text