Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012971.22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS14985-18-3
IUPAC Name(nitrooxy)(oxo)zirconio nitrate
Molecular FormulaN2O7Zr
InChI KeyJWBWFTANBHYHBC-UHFFFAOYSA-N
SMILES[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.006% max.
Zirconium dintrate oxide hydrate is used to prepare CeO2-ZrO2 mixed oxide catalyst supports. It may be used as zirconium precursor in the preparation of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and Silica- and alumina-doped nanocrystalline ceria/zirconia (Ce0.5Zr0.5O2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only