Thermo Scientific Chemicals

Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012985.22
100 g, Each
Thermo Scientific Chemicals

Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012985.22
100 g, Each
Quantity
Catalog number: 012985.22
also known as 012985-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55906-21-3
Total Metal Impurities
0.001% max.

Description

Zinc oxalate dihydrate is used as an chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text