Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013013.06
also known as 013013-06
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
Add to cart
Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number013013.06
Price (USD)/ Each
79.65
Online exclusive
87.90 
Save 8.25 (9%)
-
Add to cart
Chemical Identifiers
CAS7779-90-0
IUPAC Nametrizinc(2+) diphosphate
Molecular FormulaO8P2Zn3
InChI KeyLRXTYHSAJDENHV-UHFFFAOYSA-H
SMILES[Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only