Thermo Scientific Chemicals

Dodecacarbonyltriosmium, 99%, Thermo Scientific Chemicals

Catalog number: 013038.02
0.5 g, Each
Thermo Scientific Chemicals

Dodecacarbonyltriosmium, 99%, Thermo Scientific Chemicals

Catalog number: 013038.02
0.5 g, Each
Quantity
Catalog number: 013038.02
also known as 013038-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15696-40-9
IUPAC Name
dodecakis(methanidylidyneoxidanium) triosmium
Molecular Formula
C12O12Os3
InChI Key
VUBLMKVEIPBYME-UHFFFAOYSA-N
SMILES
[Os].[Os].[Os].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Appearance (Color)
Yellow
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
Carbon %: ≥15.6%

Description

Dodecacarbonyltriosmium is an important precursor to organo-osmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text