Thermo Scientific Chemicals

Hexacarbonylmolybdenum, 98%, Thermo Scientific Chemicals

Catalog number: 013057.22
100 g, Each
Thermo Scientific Chemicals

Hexacarbonylmolybdenum, 98%, Thermo Scientific Chemicals

Catalog number: 013057.22
100 g, Each
Quantity
Catalog number: 013057.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13939-06-5
IUPAC Name
hexakis(methanidylidyneoxidanium) molybdenum
Molecular Formula
C6MoO6
InChI Key
KMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES
[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Appearance (Color)
White to yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (special)
≥97.5% (ICP-MS)

Description

Hexacarbonylmolybdenum is widely utilized in electron beam-induced deposition technique providing an appropriate source of molybdenum atoms. It finds an application as important reagent in organometallic synthesis because of replacement of carbonyl ligands by other donor ligands. Its derivative is act as a catalyst in organic synthesis, which is used in alkyne metathesis and the Pauson-Khand reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text