Thermo Scientific Chemicals

Octacarbonyldicobalt, stab. with 1-5% hexane, Thermo Scientific Chemicals

Catalog number: 013060.88
2.5 g, Each
Thermo Scientific Chemicals

Octacarbonyldicobalt, stab. with 1-5% hexane, Thermo Scientific Chemicals

Catalog number: 013060.88
2.5 g, Each
Quantity
Catalog number: 013060.88
also known as 013060-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10210-68-1
Molecular Formula
C8Co2O8
InChI Key
ZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
Molecular Weight (g/mol)
341.95
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
Red to orange to brown
Assay (unspecified)
33 to 36 % Co

Description

As a catalyst in oxo process, and in high-purity cobalt saltsOctacarbonyldicobalt is used as a catalyst for hydroformylation. It plays an important role in promoting both the Nicholas reaction and the Pauson-Khand reaction. It acts as a reagent and catalyst in organometallic chemistry and organic synthesis. It is involved as an alkyne protecting group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text