Thermo Scientific Chemicals

Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: 013067.09
10 g, Each
Thermo Scientific Chemicals

Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: 013067.09
10 g, Each
Quantity
Catalog number: 013067.09
also known as 013067-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10028-18-9
IUPAC Name
nickel(2+) difluoride
Molecular Formula
F2Ni
InChI Key
DBJLJFTWODWSOF-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ni++]
Assay (unspecified)
96.1% min

Description

Nickel fluoride comprises the passivating surface that forms on nickel alloys, e.g. monel, which is why such materials are good to store or transport hydrogen fluoride or elemental fluorine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text