Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals
Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 013067.18
also known as 013067-18
Price (USD)/ Each
262.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
262.00
Add to cart
Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals
Catalog number013067.18
Price (USD)/ Each
262.00
-
Add to cart
Chemical Identifiers
CAS10028-18-9
IUPAC Namenickel(2+) difluoride
Molecular FormulaF2Ni
InChI KeyDBJLJFTWODWSOF-UHFFFAOYSA-L
SMILES[F-].[F-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Nickel fluoride comprises the passivating surface that forms on nickel alloys, e.g. monel, which is why such materials are good to store or transport hydrogen fluoride or elemental fluorine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only