Thermo Scientific Chemicals

Iron(III) fluoride trihydrate, Thermo Scientific Chemicals

Catalog number: 013068.30
250 g, Each
Thermo Scientific Chemicals

Iron(III) fluoride trihydrate, Thermo Scientific Chemicals

Catalog number: 013068.30
250 g, Each
Quantity
Catalog number: 013068.30
also known as 013068-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15469-38-2
Appearance (Color)
Cream to brown to red
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
Iron (Fe): ≥32.5%
Assay from Supplier's CofA
Purity: ≥97.0%

Description

The primary commercial use of iron(III) fluoride in the production of ceramics. Some cross coupling reaction are catalyzed by ferric fluoride-based compounds. Iron(III) fluoride has also been shown to catalyze chemoselective addition of cyanide to aldehydes to give the cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text