Thermo Scientific Chemicals

Manganese(III) fluoride, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 013069.09
10 g, Each
Thermo Scientific Chemicals

Manganese(III) fluoride, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 013069.09
10 g, Each
Quantity
Catalog number: 013069.09
also known as 013069-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-53-1
IUPAC Name
manganese(3+) trifluoride
Molecular Formula
F3Mn
InChI Key
SRVINXWCFNHIQZ-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Mn+3]
Total Metal Impurities
2% max.

Description

It is used as a fluorinating agent, useful for conversion of hydrocarbons into fluorocarbons. It is used in organic synthesis as Lewis acid to prepare a number of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text