Cobalt(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals
Cobalt(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 013074.22
also known as 013074-22
Price (USD)/ Each
234.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
234.00
Add to cart
Cobalt(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals
Catalog number013074.22
Price (USD)/ Each
234.00
-
Add to cart
Chemical Identifiers
CAS10026-17-2
IUPAC Nameλ²-cobalt(2+) difluoride
Molecular FormulaCoF2
InChI KeyYCYBZKSMUPTWEE-UHFFFAOYSA-L
SMILES[F-].[F-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Cobalt(II) fluoride can be used as a catalyst to alloy metals. It is also used for optical deposition, of which it tremendously improves optical quality. The compound may be used in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only