Thermo Scientific Chemicals

Bismuth(III) fluoride, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 013075.22
100 g, Each
Thermo Scientific Chemicals

Bismuth(III) fluoride, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 013075.22
100 g, Each
Quantity
Catalog number: 013075.22
also known as 013075-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-61-3
IUPAC Name
bismuth(3+) trifluoride
Molecular Formula
BiF3
InChI Key
BRCWHGIUHLWZBK-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Bi+3]
Assay (unspecified)
98.6% min

Description

It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text