Thermo Scientific Chemicals

Titanium(III) fluoride, Thermo Scientific Chemicals

Catalog number: 013079.09
10 g, Each
Thermo Scientific Chemicals

Titanium(III) fluoride, Thermo Scientific Chemicals

Catalog number: 013079.09
10 g, Each
Quantity
Catalog number: 013079.09
also known as 013079-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13470-08-1
IUPAC Name
titanium(3+) trifluoride
Molecular Formula
F3Ti
InChI Key
NLPMQGKZYAYAFE-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Ti+3]
Assay (unspecified)
53.26% F min

Description

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text