Thermo Scientific Chemicals

Titanium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 013087.18
50 g, Each
Thermo Scientific Chemicals

Titanium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 013087.18
50 g, Each
Quantity
Catalog number: 013087.18
also known as 013087-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12067-57-1
IUPAC Name
trisilicon pentatitanium
Molecular Formula
Si3Ti5
InChI Key
GNKTZDSRQHMHLZ-UHFFFAOYSA-N
SMILES
[Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti]
Total Metal Impurities
0.5% max.

Description

It is used as source, gate and drain contacts and local interconnections in CMOS integrated circuits. it is used in the C54 phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text