Thermo Scientific Chemicals

Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 013093.09
10 g, Each
Thermo Scientific Chemicals

Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 013093.09
10 g, Each
Quantity
Catalog number: 013093.09
also known as 013093-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12033-62-4
Total Metal Impurities
0.5% max.

Description

Tantalum nitride is used to create barrier or glue layers between copper, or other conductive metals, and dielectric insulator films such as thermal oxides. These films are deposited on top of silicon wafers during the manufacture of integrated circuits, to create thin film surface mount resistors and has other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text