Thermo Scientific Chemicals

Nickel boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013110.22
100 g, Each
Thermo Scientific Chemicals

Nickel boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013110.22
100 g, Each
Quantity
Catalog number: 013110.22
also known as 013110-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-00-0
IUPAC Name
nickelioborane
Molecular Formula
BNi
InChI Key
QDWJUBJKEHXSMT-UHFFFAOYSA-N
SMILES
B#[Ni]
Total Metal Impurities
1% max.

Description

Nickel boride is an efficient catalyst and reducing agent. It is used as a heterogeneous hydrogenation catalyst. It can also be used to cleave thioacetals. It is used in a protection-deprotection scheme for triterpenoid ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text