Thermo Scientific Chemicals

Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013126.18
50 g, Each
Thermo Scientific Chemicals

Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013126.18
50 g, Each
Quantity
Catalog number: 013126.18
also known as 013126-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12068-69-8
IUPAC Name
dibismuth(3+) triselandiide
Molecular Formula
Bi2Se3
InChI Key
FBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES
[Se--].[Se--].[Se--].[Bi+3].[Bi+3]
Form
Pieces, 1-6 mm
Total Metal Impurities
≤0.005%

Description

In semiconductor research, multilayer thin-film, magnetoresistant film.Bismuth(III) selenide is used as a semiconductor and thermoelectric material. It is also used in semiconductor research. Further, it finds applications magnetoresistant film and semiconductor research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text