Manganese(IV) telluride, 99.9% (metals basis), Thermo Scientific Chemicals
Manganese(IV) telluride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(IV) telluride, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 013134.18
also known as 013134-18
Price (USD)/ Each
364.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
364.00
Add to cart
Manganese(IV) telluride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number013134.18
Price (USD)/ Each
364.00
-
Add to cart
Chemical Identifiers
CAS12032-89-2
IUPAC Nameditellanylidenemanganese
Molecular FormulaMnTe2
InChI KeyKDJKBQYOEWVJGP-UHFFFAOYSA-N
SMILES[Te]=[Mn]=[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
It is used as an organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only