Thermo Scientific Chemicals
Iron(II) titanium oxide, 99.8+% (metals basis), Thermo Scientific Chemicals
Catalog number: 013137.30
250 g, Each
Thermo Scientific Chemicals
Iron(II) titanium oxide, 99.8+% (metals basis), Thermo Scientific Chemicals
Catalog number: 013137.30
250 g, Each
Quantity
Catalog number: 013137.30
also known as 013137-30
Price (USD)
Quantity
-
Have Questions?
Chemical Identifiers
CAS
12022-71-8
Total Metal Impurities
0.2% max.
Description
Iron(II) titanium oxide is used as a chief source of titanium. It is also used to prepare titanium dioxide by reacting sulfuric acid to get titanium oxygen sulfate followed by hydrolysis.