Copper(I) telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Copper(I) telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(I) telluride, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 013140.14
also known as 013140-14
Price (USD)/ Each
292.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
292.00
Add to cart
Copper(I) telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number013140.14
Price (USD)/ Each
292.00
-
Add to cart
Chemical Identifiers
CAS12019-52-2
IUPAC Name(cupriotellanyl)copper
Molecular FormulaCu2Te
InChI KeyMZEWONGNQNXVKA-UHFFFAOYSA-N
SMILES[Cu][Te][Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only