Thermo Scientific Chemicals

Nitronium hexafluorophosphate, Thermo Scientific Chemicals

Catalog number: 013151.06
5 g, Each
Thermo Scientific Chemicals

Nitronium hexafluorophosphate, Thermo Scientific Chemicals

Catalog number: 013151.06
5 g, Each
Quantity
Catalog number: 013151.06
also known as 013151-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19200-21-6
IUPAC Name
dioxoamine; hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6HNO2P
InChI Key
WCPUJULSPZCGOX-UHFFFAOYSA-N
SMILES
[O-][NH+]=O.F[P-](F)(F)(F)(F)F
Assay (unspecified)
96% min

Description

Nitronium hexafluorophosphate is a nitrating reagent used in the nitration of alkanes such as methane, ethane, propane, n-butane, isobutane, neopentane and cyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text