Thermo Scientific Chemicals

Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013154.22
100 g, Each
Thermo Scientific Chemicals

Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013154.22
100 g, Each
Quantity
Catalog number: 013154.22
also known as 013154-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207801-29-4
IUPAC Name
yttrium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21O6Y
InChI Key
CKIVBSKECCTIAG-UHFFFAOYSA-N
SMILES
[Y+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Yttrium(III) 2,4-pentanedionate hydrate acts as a precursor with europium nitrate and used for the preparation of eurobium-doped yttrium orthovanadate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text