Thermo Scientific Chemicals

Ytterbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013159.09
10 g, Each
Thermo Scientific Chemicals

Ytterbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013159.09
10 g, Each
Quantity
Catalog number: 013159.09
also known as 013159-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14284-98-1
IUPAC Name
ytterbium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21O6Yb
InChI Key
JKJCKIFLFXFUKJ-UHFFFAOYSA-N
SMILES
[Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Ytterbium(III) 2,4-pentanedionate is used as a catalyst for the preparation of aliphatic oligocarbonate polyols. It takes part in transesterification reactions. It is used for the preparation of polyesters or polyacrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text