Neodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Neodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 013185.14
also known as 013185-14
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
Neodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number013185.14
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS13477-91-3
IUPAC Namedineodymium(3+) octahydrate trisulfate
Molecular FormulaH16Nd2O20S3
InChI KeyIOUARGWNXMWCLD-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Nd+3].[Nd+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Neodymium(III) sulfate is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only