Thermo Scientific Chemicals

Lutetium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013189.06
5 g, Each
Thermo Scientific Chemicals

Lutetium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013189.06
5 g, Each
Quantity
Catalog number: 013189.06
also known as 013189-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17966-84-6
IUPAC Name
lutetium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21LuO6
InChI Key
HXFHOORFYPCGNF-UHFFFAOYSA-N
SMILES
[Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lutetium(III) 2,4-pentanedionate is involved in refluxing 1,2,4-trichlorobenzene. It is also used as catalyst of organic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text