Thermo Scientific Chemicals

Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013197.18
50 g, Each
Thermo Scientific Chemicals

Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013197.18
50 g, Each
Quantity
Catalog number: 013197.18
also known as 013197-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54451-24-0
IUPAC Name
dilanthanum(3+) tricarbonate
Molecular Formula
C3La2O9
InChI Key
NZPIUJUFIFZSPW-UHFFFAOYSA-H
SMILES
[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Lanthanum(III) carbonate hydrate is used as a starting material for the preparation of lanthanum strontium manganite, which is used in solid oxide fuel cell applications. It is also involved in the preparation of high-temperature superconductors. It finds application in medicine as a phosphate binder. Further, it is used in the treatment of hyperphosphatemia. In addition to this, it is used for the tinting of glass, water treatment and as a catalyst for hydrocarbon cracking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text