Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 013198.09
also known as 013198-09
Price (USD)/ Each
31.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
31.10
Add to cart
Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number013198.09
Price (USD)/ Each
31.10
-
Add to cart
Chemical Identifiers
CAS224183-16-8
IUPAC Namelanthanum(3+) tribromide
Molecular FormulaBr3La
InChI KeyXKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only